Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF5 Mouse anti-Human, Clone:6D6H11, Abnova™

Catalog No. 89334460 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-460 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-460 Supplier Abnova Corporation Supplier No. MAB17817
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human EIF5.

Specifications

Antigen EIF5
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6D6H11
Conjugate Unconjugated
Description eukaryotic translation initiation factor 5
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry (1:50-1:250)
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene EIF5
Gene Alias EIF-5, EIF-5A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-300 of human EIF5 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 1983
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.