Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528488 Supplier Invitrogen™ Supplier No. PA528488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLaS3, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (93%), Xenopus laevis (93%), Chicken (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Eif5
Gene Accession No. P55010
Gene Alias 2810011H21Rik; D12Ertd549e; EIF5; EIF-5; EIF-5A; eukaryotic initiation factor 5 (eIF-5); eukaryotic translation initiation factor 5
Gene Symbols Eif5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EIF5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.