Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF5A Antibody (4D1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259403APC
This item is not returnable.
View return policy
Description
eIF5A Monoclonal antibody specifically detects eIF5A in Human samples. It is validated for Flow CytometrySpecifications
eIF5A | |
Monoclonal | |
APC | |
eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
Recombinant human eIF5A (1-154aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Cell Biology | |
1984 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
4D1 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction