Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF5A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | eIF5A |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
eIF5A Polyclonal specifically detects eIF5A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
eIF5A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% with 0.02% Sodium Azide | |
eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
EIF5A | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
P63241 | |
1984 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MCGTGGTDSKTRRPPHRASFLKRLE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title