Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563944 Supplier Invitrogen™ Supplier No. PA563944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCGTGGTDSK TRRPPHRASF LKRLE Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.

mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. With syntenin SDCBP, functions as a regulator of TP53/p53 and TP53/p53-dependent apoptosis. Regulates also TNF-alpha-mediated apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation. Also described as a cellular cofactor of human T-cell leukemia virus type I (HTLV-1) Rex protein and of human immunodeficiency virus type 1 (HIV-1) Rev protein, essential for mRNA export of retroviral transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF5A
Gene Accession No. P63241
Gene Alias AA410058; D19Wsu54e; Eif4d; eIF-4D; Eif5a; eIF-5A; EIF5A1; eIF-5A1; eIF-5A-1; eIF5AI; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; eukaryotic translation initiation factor 5A; eukaryotic translation initiation factor 5A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A
Gene Symbols EIF5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eIF5A. Recombinant protein control fragment (Product #RP-103395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.