Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF5A2 Antibody (OTI6G7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272056H
Description
eIF5A2 Monoclonal specifically detects eIF5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
eIF5A2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
EIF5A2 | |
Full length recombinant protein of human eIF5A2 (NP_065123) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6G7 | |
Western Blot, Immunohistochemistry | |
eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2 | |
Mouse | |
Affinity Purified | |
Cancer, Tumor Suppressors | |
56648.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction