Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF5A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587136
Description
Purity is > 95% by SDS-PAGE.
mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. Functions as a regulator of apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation.
Specifications
eIF5A2 | |
Polyclonal | |
Unconjugated | |
EIF5A2 | |
9630038B20; EIF5A2; eIF-5A2; eIF-5A-2; eIF5AII; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; eukaryotic translation initiation factor 5A2; Eukaryotic translation initiation factor 5A-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
208691, 310261, 56648 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BGY2, Q9GZV4 | |
EIF5A2 | |
Recombinant fusion protein of human EIF5A2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction