Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120894 Supplier Invitrogen™ Supplier No. PA5120894
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, SKOV3, SW480, THP-1, Mouse lung, Mouse brain, Mouse spleen, Rat lung. The target is usually found in the following locations: Cytoplasm, Cytoplasmic side, Endoplasmic reticulum membrane, Nucleus, Peripheral membrane protein, nuclear pore complex. Immunogen sequence: MADEIDFTTG DAGASSTYPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDYQ LICIQDGYLS LLTETGEVRE DLKLPEGELG KEIEGKYNAG EDVQVSVMCA MSEEYAVAIK PCK.

mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. Functions as a regulator of apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EIF5A2
Gene Accession No. Q8BGY2, Q9GZV4
Gene Alias 9630038B20; EIF5A2; eIF-5A2; eIF-5A-2; eIF5AII; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; eukaryotic translation initiation factor 5A2; Eukaryotic translation initiation factor 5A-2
Gene Symbols EIF5A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-153 of human EIF5A2 (NP_065123.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208691, 310261, 56648
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.