Learn More
Invitrogen™ EIF5AL1 Polyclonal Antibody

Description
EIF5AL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | EIF5AL1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EIF5AL1 |
| Gene Accession No. | Q6IS14 |
| Gene Alias | bA342M3.3; eIF-5A-1-like; eIF-5A1-like; EIF5AL1; EIF5AP1; Eukaryotic initiation factor 5A isoform 1-like; eukaryotic translation initiation factor 5A pseudogene 1; eukaryotic translation initiation factor 5A-1-like; eukaryotic translation initiation factor 5A-like 1 |
| Gene Symbols | EIF5AL1 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.