Learn More
eIF5B Monoclonal Antibody (3F9)

Description
eIF5B Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | eIF5B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3F9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EIF5B |
| Gene Accession No. | O60841 |
| Gene Alias | A030003E17Rik; Annexin V-binding protein ABP-7; BC018347; DKFZp434I036; eIF 5B; Eif5b; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; If2; KIAA0741; translation initiation factor IF2; Translation initiation factor IF-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.