Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565258 Supplier Invitrogen™ Supplier No. PA565258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGKLLTKSQRE ARARAEATLK LLQAQGVEVP SKDSLPKKRP IYEDKKRKKI PQQLESKEVS ESMELCAAVE VMEQGVPEKE Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.

Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF5B
Gene Accession No. O60841
Gene Alias A030003E17Rik; Annexin V-binding protein ABP-7; BC018347; DKFZp434I036; eIF 5B; Eif5b; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; If2; KIAA0741; translation initiation factor IF2; Translation initiation factor IF-2
Gene Symbols EIF5B
Host Species Rabbit
Immunogen Recombinant Human eIF5B. Recombinant protein control fragment (Product #RP-104713).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.