Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF6 Monoclonal Antibody (A6A3)
GREENER_CHOICE

Catalog No. PIMA542548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542548 Supplier Invitrogen™ Supplier No. MA542548
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: Hela cell lysate, CRC cell lysate, NIH/3T3 cell lysate, PC-12 cell lysate, human esophagus tissue, human ovarian carcinoma tissue, mouse colon tissue. Subcellular Location: Nucleolus, Cytoplasm.

Hemidesmosomes are structures which link the basal lamina to the intermediate filament cytoskeleton. An important functional component of hemidesmosomes is the integrin beta-4 subunit (ITGB4), a protein containing two fibronectin type III domains. The protein encoded by this gene binds to the fibronectin type III domains of ITGB4 and may help link ITGB4 to the intermediate filament cytoskeleton. The encoded protein, which is insoluble and found both in the nucleus and in the cytoplasm, can function as a translation initiation factor and prevent the association of the 40S and 60S ribosomal subunits. Multiple transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6A3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Eif6
Gene Accession No. O55135, P56537, Q3KRD8
Gene Alias 1110004P11; AA408895; b(2)gcn; B(2)GCN homolog; B4 integrin interactor; CAB; EIF3A; Eif6; eIF-6; eIF-6 {ECO:0000255; eukaryotic translation initiation factor 3A; eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255; HAMAP-Rule:MF_03132}; imc-415; integrin beta 4 binding protein; Itgb4bp; OK/SW-cl.27; p27 beta-4 integrin-binding protein; p27(BBP); p27BBP
Gene Symbols Eif6
Host Species Mouse
Immunogen Recombinant protein within human eIF-6 aa 1-200.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16418, 305506, 3692
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.