Learn More
Invitrogen™ eIF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579197
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human K562 whole cell, human 293T whole cell, human HepG2 whole cell, rat liver tissue, mouse liver tissue, mouse RAW2647 whole cell. IHC: Mouse Intestine tissue, Rat Testis tissue, Human Intestinal Cancer tissue. ICC/IF: SMMC-7721 Cell, SW480 Cell.
Hemidesmosomes are structures which link the basal lamina to the intermediate filament cytoskeleton. An important functional component of hemidesmosomes is the integrin beta-4 subunit (ITGB4), a protein containing two fibronectin type III domains. The protein encoded by this gene binds to the fibronectin type III domains of ITGB4 and may help link ITGB4 to the intermediate filament cytoskeleton. The encoded protein, which is insoluble and found both in the nucleus and in the cytoplasm, can function as a translation initiation factor and prevent the association of the 40S and 60S ribosomal subunits. Multiple transcript variants encoding two different isoforms have been found for this gene.
Specifications
eIF6 | |
Polyclonal | |
Unconjugated | |
Eif6 | |
1110004P11; AA408895; b(2)gcn; B(2)GCN homolog; B4 integrin interactor; CAB; EIF3A; Eif6; eIF-6; eIF-6 {ECO:0000255; eukaryotic translation initiation factor 3A; eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255; HAMAP-Rule:MF_03132}; imc-415; integrin beta 4 binding protein; Itgb4bp; OK/SW-cl.27; p27 beta-4 integrin-binding protein; p27(BBP); p27BBP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16418, 305506, 3692 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O55135, P56537, Q3KRD8 | |
Eif6 | |
E.coli-derived human EIF6 recombinant protein (Position: N66-T210). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.