Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144402
Description
EIN2 Polyclonal Antibody for Western Blot, ELISA
Central factor in signaling pathways regulated by ethylene (ET) and involved in various processes including development, plant defense, senescence, nucleotide sugar flux, and tropisms (PubMed:27694846, PubMed:28874528). Necessary for ethylene-mediated gene regulation, and for the induction of some genes by ozone. Acts downstream of ET receptors, and upstream of ethylene regulated transcription factors. Required for cytokinin-mediated processes. Seems to be implicated in cross-talk between ET, jasmonate and other pathways. Probably not involved in iron uptake (PubMed:10381874, PubMed:12953109, PubMed:14973160, PubMed:15010611, PubMed:15272873, PubMed:7770519, PubMed:9351240). Has a short half-life and undergoes rapid proteasome-mediated turnover in the absence of ethylene (PubMed:19196655). Required for ethylene-induced EIN3 stabilization via proteasomal degradation of EBF1/EBF2 proteins (PubMed:20647342). Regulates the leaf senescence induced by methyl jasmonate, ethylene and abscisic acid (Ref.17, PubMed:15010611). Required during salt stress to confer resistance (PubMed:21631530).
Specifications
| EIN2 | |
| Polyclonal | |
| Unconjugated | |
| EIN2 | |
| CKR1; ORE3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 831889 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.103 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9S814 | |
| EIN2 | |
| Recombinant Arabidopsis thaliana EIN2 protein (747-927aa). | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction