Description
- Small clast size indicates deposition in low-energy environment such as shallow lake or slow-moving river
- For use in sedimentary rock identification exercises due to its diagnostic flat, fissile rock structure
- Correlates well to Next Generation Science Standards Evidence Statements
- Small clast size indicates that though the grains had been eroded, rock formed in low energy environment
Specifications
Specifications
| Product Type | Sedimentary Rock |
| For Use With (Application) | Sedimentary rock identification exercises due to its diagnostic flat, fissile rock structure |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Argillaceous Shale |