Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Mixing Box
SCI_ED

Catalog No. S12965
Click to view available options
:
Colour Mixing Box

Light: Standard Lighting

  • Optical exploration kit includes activities to engage students in both additive and subtractive color mixing
  • With the color mixing box, students can control the brightness of each color light (Blue, Red, and Green) to discover how each of two or three colors add to make the rest of the colors in the rainbow
  • Also included is a set of 24 water color based paints for students to compare the subtractive color mixing process with that of additive process in the light box
  • Powered by 3 AA batteries, it's perfect for desktop laboratory experimentation
  • Color mixing box measures 10 x 7 x 5.5 inches
  • Batteries included

Specifications

Product Type Mixing Box
Includes Set of 24 water color based paints, batteries
Width (English) 7.9 in.
Width (Metric) 20 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.