Learn More
Eisco™ pH Sensor
Allows students to easily measure pH of pure chemicals and dynamic chemical reactions.
Supplier: Eisco™ ESP105
Requires : Requires Eisco Sensors Base Unit, available separately.
Description
Specifications:- Range: 0 to 14pH
- Resolution: 0.01 pH
- Sample Rate: 100 S/sec
- Analysis of chemical reactions in real time, analysis of static chemicals, environmental monitoring, and water quality testing
Specifications
Sensor, pH | |
Analysis of chemical reactions in real time, analysis of static chemicals, environmental monitoring, and water quality testing |