Learn More
Eisco™ Scalpel Blades
SCI_ED
Catalog No. S95938B Shop All Eisco Products
Change view
Click to view available options
Size:
# 10
# 11
# 22
# 23
# 24
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S95938B -
S95937A # 10
S95937B # 11
S95938A -
S95938C # 22
S95938D # 23
S24469 # 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S95938B Supplier Eisco™ Supplier No. BI0184B21
Add to Cart
Add to Cart
Quality blades, individually packed in aluminum foil.
  • Blades 10 and 11 are suitable for use with handle No. 3 (S95937)
  • Blades 20 through 23 are suitable for handle No. 4 (S95938)
Order Info
Sold as pack of 10.