Learn More
Eisco™ Scalpel Blades
Quality blades, individually packed in aluminum foil.
$1.59 - $10.87
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
S95937A
![]() |
Eisco™
BI0184A10 |
# 10 |
Pack of 10 for $7.95
|
![]() |
|||||
S95937B
![]() |
Eisco™
BI0184A11 |
# 11 |
Pack of 10 for $7.95
|
|
|||||
S95938A
![]() |
Eisco™
BI0184B20 |
Pack of 10 for $1.59
|
|
||||||
S95938B
![]() |
Eisco™
BI0184B21 |
Pack of 10 for $10.87
|
|
||||||
S95938C
![]() |
Eisco™
BI0184B22 |
# 22 |
Pack of 10 for $7.95
|
|
|||||
S95938D
![]() |
Eisco™
BI0184B23 |
# 23 |
Pack of 10 for $7.47
|
|
|||||
S24469
![]() |
Eisco™
BI0184B24 |
Each for $2.44
|
|
||||||
Description
- Blades 10 and 11 are suitable for use with handle No. 3 (S95937)
- Blades 20 through 23 are suitable for handle No. 4 (S95938)
Specifications
Scalpel Blade |