Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Visual Scientifics Pendulum
SCI_ED

Catalog No. s06900
Click to view available options
Grade:
9 to 12

Includes

Extensive instructions to assist with lesson planning, student activities and experimental procedures; contains pendulum back board, mounting rod, string, pendulum bob and pendulum stopper

Inquiry-based activity to discover the period of a simple pendulum.

Technology allows students to study pendulums and frequency in different dimensions and speeds.

  • Main Lab: What is the period of a pendulum?
  • Extension One: energy conservation
  • Extension Two: applying technology to track energy conversion
  • Covers 10 Next Generation Science Standards Concepts
  • Covers eight Common Core Standards
  • Approximately three hours of class/lab time material
  • 20 pages in total
  • Student workbook is a guide for students to fill out as they work through the main lab and lab extensions
  • General Theory Section lays out the theory and mathematics behind the lesson plan
  • Experimental Teaching Guide helps teachers prepare lesson plan and lead inquiry or step-based activities
  • All labs and extensions include recommenations for video analysis setups

Order Info

For use with Physical Science System Base (S06893).

Specifications

Grade 9 to 12
Format Kit
Product Type Visual Scientifics Pendulum
Includes Extensive instructions to assist with lesson planning, student activities and experimental procedures, contains pendulum back board, mounting rod, string, pendulum bob and pendulum stopper
Languages English
For Use With (Application) Inquiry based activity to discover the period of a simple pendulum
Subject Physics
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.