Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier EKG Sensor
SDPSCI_ED

Catalog No. s16142nd Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16142ND High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16142ND Supplier Vernier Supplier No. EKGBTA
Only null left
Add to Cart
Add to Cart

Requires

An interface and software, sold separately

Measures electrical signals produced during muscle contractions.

  • Measures cardiac electrical potential waveforms (voltages produced during the contraction of the heart)
  • Can be used to make standard 3-lead EKG tracings to record electrical activity in the heart, or to collect surface EMG recordings to study contractions in muscles in your arm, leg or jaw

 

Specifications:

  • Offset: ∽1.00 V (±0.3V)
  • Gain: 1mV body potential / 1V sensor output
  • Includes user manual

 

Data Collection:

This sensor can be used with the following interfaces to collect data:
  • Vernier LabQuest™ 2 or original LabQuest™ as a standalone device or with a computer
  • Vernier LabQuest™ Mini with a computer
  • Vernier LabPro™ with a computer or TI graphing calculator
  • Vernier Go!™Link1
  • Vernier EasyLink™
  • Vernier SensorDAQ™
  • CBL 2™
  • TI-Nspire™ Lab Cradle

Specifications

Includes Pack of 100 disposable electrodes and user manual
Grade High School and College
For Use With (Application) Measure electrical signals produced during muscle contractions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.