Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EKLF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518031
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518031 Supplier Invitrogen™ Supplier No. PA518031
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EKLF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Klf1
Gene Accession No. Q13351
Gene Alias CDAN4; Eklf; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1
Gene Symbols Klf1
Host Species Goat
Immunogen Synthetic peptide sequence (ATAETALPSISTLT) corresponding to the N-terminus amino acids of KLF1 (aa 2-16).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.