Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EKLF Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA584089
Description
Immunogen sequence: LALQPVYPGP GAGSSGGYFP RTGLSVPAAS GAPYGLLSGY PAMYPAPQYQ GHFQLFRGLQ GPAPGPATSP SFLSCLGPG
This gene encodes a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the domit Inblood phenotype.
Specifications
EKLF | |
Polyclonal | |
Unconjugated | |
KLF1 | |
EKLF | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13351 | |
KLF1 | |
Recombit protein corresponding to Human KLF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction