Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELA3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565016 Supplier Invitrogen™ Supplier No. PA565016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTCGGSLIAPD WVVTAGHCIS RDLTYQVVLG EYNLAVKEGP EQVIPINSEE LFVHPLWNRS CVACGNDIAL IKLSRSAQLG DAVQLASLPP AGDIL Highest antigen sequence identity to the following orthologs - mouse 78%, rat 77%.

Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3A has little elastolytic activity. Like most of the human elastases, elastase 3A is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein, it has a digestive function in the intestine. Elastase 3A preferentially cleaves proteins after alanine residues. Elastase 3A may also function in the intestinal transport and metabolism of cholesterol. Both elastase 3A and elastase 3B have been referred to as protease E and as elastase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELA3A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CELA3A
Gene Accession No. P09093
Gene Alias CELA3A; chymotrypsin like elastase family member 3A; chymotrypsin-like elastase family member 3A; chymotrypsin-like elastase family, member 3A; ELA3; ELA3A; elastase 1; elastase 3A, pancreatic; elastase IIIA; Elastase-3A; Gm13011; OTTMUSG00000009877; protease E; uncharacterized protein LOC242711
Gene Symbols CELA3A
Host Species Rabbit
Immunogen Recombinant Human ELA3A. Recombinant protein control fragment (Product #RP-105608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.