Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELA3B Rabbit anti-Human, Unconjugated, Clone: CELA3B/2809R, Abnova™

Catalog No. 89337838 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-838 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-838 Supplier Abnova Corporation Supplier No. RAB00413
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

ELA3B Monoclonal antibody specifically detects ELA3B in Human samples. It is validated for IHC-P.

Specifications

Antigen ELA3B
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CELA3B/2809R
Conjugate Unconjugated
Description elastase 3B, pancreatic
Gene ELA3B
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acids 82-238 of human ELA3B
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 23436
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.