Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216313
Description
ELAC1 Polyclonal specifically detects ELAC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ELAC1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
D29elaC (E. coli) homolog 1, Deleted in Ma29, EC 3.1.26.11, elaC homolog 1 (E. coli), ElaC homolog protein 1, FLJ59261, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNA 3' processing endoribonuclease, tRNA Z (short form), tRNase Z 1, tRNase ZS, zinc phosphodiesterase ELAC protein 1 | |
Rabbit | |
40 kDa | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
55520 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q9H777 | |
ELAC1 | |
Recombinant protein encompassing a sequence within the center region of human ELAC1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (89%), Rat (89%), Bovine (92%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction