Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531106 Supplier Invitrogen™ Supplier No. PA531106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (89%), Rat (89%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Zinc phosphodiesterase, which displays some tRNA 3'-processing endonuclease activity. Probably involved in tRNA maturation, by removing a 3'-trailer from precursor tRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Elac1
Gene Accession No. Q9H777
Gene Alias 2610018O07Rik; 8430417G19Rik; D29; Deleted in Ma29; elaC homolog 1; elaC homolog 1 (E. coli); elaC homolog protein 1; elaC ribonuclease Z 1; Elac1; Ribonuclease Z 1; RNase Z 1; RNaseZ(S); tRNA 3 endonuclease 1; tRNA 3' processing endoribonuclease; tRNA Z (short form); tRNase Z 1; tRNase ZS; zinc phosphodiesterase ELAC protein 1
Gene Symbols Elac1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ELAC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55520
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.