Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELAC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566844
Description
Immunogen sequence: IWRTMELSHTE LVFHYVVHEL VPTADQCPAE ELKEFAHVNR ADSPPKEEQG RTILLDSEEN SYLLFDDEQF VVKAFRLFHR IPSFGF Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.
Zinc phosphodiesterase, which displays some tRNA 3'-processing endonuclease activity. Probably involved in tRNA maturation, by removing a 3'-trailer from precursor tRNA.
Specifications
ELAC1 | |
Polyclonal | |
Unconjugated | |
Elac1 | |
2610018O07Rik; 8430417G19Rik; D29; Deleted in Ma29; elaC homolog 1; elaC homolog 1 (E. coli); elaC homolog protein 1; elaC ribonuclease Z 1; Elac1; Ribonuclease Z 1; RNase Z 1; RNaseZ(S); tRNA 3 endonuclease 1; tRNA 3' processing endoribonuclease; tRNA Z (short form); tRNase Z 1; tRNase ZS; zinc phosphodiesterase ELAC protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H777 | |
Elac1 | |
Recombinant Human ELAC1. Recombinant protein control fragment (Product #RP-107498). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction