Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735410 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5410 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5410 Supplier Proteintech Group Inc Supplier No. 266391AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zinc phosphodiesterase, which displays some tRNA 3′-processing endonuclease activity. Probably involved in tRNA maturation, by removing a 3′-trailer from precursor tRNA.

Specifications

Antigen ELAC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ELAC1
Gene Accession No. Q8VEB6, Q9H777
Gene Alias D29, Deleted in Ma29, elaC homolog 1 (E. coli), ElaC homolog protein 1, ELAC1, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNase Z 1
Gene Symbols ELAC1
Host Species Rabbit
Immunogen ELAC1 Fusion Protein Ag23939
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114615, 55520
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.