Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541987 Supplier Invitrogen™ Supplier No. PA541987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ITCNPEEFIV EALQLPNFQQ SVQEYRRSAQ DGPAPAEKRS QYPEIIFLGT Sequence homology: Dog: 100%; Human: 100%; Mouse: 83%; Rat: 83%.

ELAC2 encodes a protein that has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Elac2
Gene Accession No. Q9BQ52
Gene Alias 1110017O07Rik; COXPD17; D11Wsu80e; elaC homolog 2; elaC homolog 2 (E. coli); elaC homolog protein 2; elaC ribonuclease Z 2; ELAC2; elaC-like protein 2; ELC2; heredity prostate cancer protein 2; HPC2; putative prostate cancer susceptibility protein HPC2/ELAC2; ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; tRNase Z(L); zinc phosphodiesterase ELAC protein 2
Gene Symbols Elac2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ELAC2 (aa 439-488).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60528
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.