Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554016 Supplier Invitrogen™ Supplier No. PA554016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WMERFGPDTQ HLVLNENCAS VHNLRSHKIQ TQLNLIHPDI FPLLTSFRCK KEGPTLSVPM VQGECLLKYQ LRPRREWQRD AIITCNPEEF IVEALQLPNF QQSVQEYR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

ELAC2 encodes a protein that has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Elac2
Gene Accession No. Q9BQ52
Gene Alias 1110017O07Rik; COXPD17; D11Wsu80e; elaC homolog 2; elaC homolog 2 (E. coli); elaC homolog protein 2; elaC ribonuclease Z 2; ELAC2; elaC-like protein 2; ELC2; heredity prostate cancer protein 2; HPC2; putative prostate cancer susceptibility protein HPC2/ELAC2; ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; tRNase Z(L); zinc phosphodiesterase ELAC protein 2
Gene Symbols Elac2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELAC2. Recombinant protein control fragment (Product #RP-92519).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.