Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELAC2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568644
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8644 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8644 Supplier Bethyl Laboratories, Inc Supplier No. A304775AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3′ trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009].

Specifications

Antigen ELAC2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ELAC2
Gene Accession No. Q80Y81, Q9BQ52
Gene Alias ELC2, HPC2, RNase Z 2, elaC homolog protein 2, heredity prostate cancer protein 2, putative prostate cancer susceptibility protein HPC2/ELAC2, ribonuclease Z 2, tRNA 3 endonuclease 2, tRNase Z (long form), tRNase Z 2, zinc phosphodiesterase ELAC protein 2
Gene Symbols ELAC2
Host Species Rabbit
Immunogen Between 776 and 826
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60528, 68626
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.