Learn More
Elaidic acid, 98%
Description
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 41°C to 45°C |
| Boiling Point | 288°C (100 mm) |
| Flash Point | >110°C (230°F) |
| Assay Percent Range | 98% |
| Quantity | 1 g |
| Beilstein | 1726543 |
| Merck Index | 14,3533 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 282.47 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 204-006-6 |
| RTECSNumber | JX6125000 |
| TSCA | No |
| storageNote1 | Air sensitive |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.