Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elaidic acid, 98%

Catalog No. p-7022187
Click to view available options
Quantity:
1 g
5 g
25 g

Store cold | CAS: 112-79-8 | C18H34O2 | 282.468 g/mol

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00063954
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O

Specifications

Melting Point 41°C to 45°C
Boiling Point 288°C (100 mm)
Flash Point >110°C (230°F)
Assay Percent Range 98%
Quantity 1 g
Beilstein 1726543
Merck Index 14,3533
Solubility Information Insoluble in water.
Formula Weight 282.47
Percent Purity 98%
Chemical Name or Material Elaidic Acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-006-6
RTECSNumber JX6125000
TSCA No
storageNote1 Air sensitive
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.