Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elaidic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187729 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Packaging Poly Bottle
CAS 112-79-8
Flash Point >113°C
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOH
MDL Number MFCD00063954
Melting Point 42°C to 44°C
Boiling Point 288°C (100 mmHg)
Synonym trans-9-Octadecenoic acid; trans-Oleic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 282.46
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.