Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastase Antibody (CELA3B/1257), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253246200UG
Description
Elastase Monoclonal antibody specifically detects Elastase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Elastase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Mouse | |
| 17 kDa | |
| 200 μg | |
| Cancer, Neuroscience | |
| 23436 | |
| Store at 4C. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CELA3B/1257 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| CELA3B | |
| Recombinant human CELA3B protein fragment (aa82-238) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction