Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastase Antibody (CELA3B/1758), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308863R
Description
Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Western Blot, ELISASpecifications
Elastase | |
Monoclonal | |
DyLight 550 | |
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
Recombinant human Elastase protein fragment (aa82-238) (exact sequence is proprietary) (Uniprot: P08861) | |
100 μg | |
Cancer, Neuroscience | |
23436 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
CELA3B/1758 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction