Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastase Antibody (CELA3B/1811), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Elastase |
---|---|
Clone | CELA3B/1811 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25328420
![]() |
Novus Biologicals
NBP25328420UG |
20 μg |
Each for $268.00
|
|
|||||
NB253284100
![]() |
Novus Biologicals
NBP253284100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253284200
![]() |
Novus Biologicals
NBP253284200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Elastase | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Cancer, Neuroscience | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
23436 | |
Recombinant human CELA3B protein fragment (aa82-238) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
CELA3B/1811 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
CELA3B | |
IgG2b κ | |
Protein A or G purified | |
17 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title