Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Elastase (Porcine)

Catalog No. 89943132
Click to view available options
Quantity:
5 mg

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 25.9kDa
Name Elastase (Porcine)
Preparation Method Native protein chromatographically purified from Porcine Pancreas
Quantity 5 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name ELASTASE
Assays Elastase Assay
Biological Activity ≥8 units per mg protein. One Unit cleaves one micromole of N-succinyl-L-alanyl-L-alanyl-L-alanine-p-nitroanilide per minute at 25°C, pH 8.0.
Species Pig
Protein Tag None
Expression System Native protein chromatographically purified from Porcine Pancreas
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.