Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase, Porcine Pancreas, PROMO, Thermo Scientific Chemicals

Catalog No. J61874MCPM
Click to view available options
Quantity:
100 mg

Elastase, porcine pancreas is a single polypeptide chain of 240 amino acid residues isolated from the porcine pancreas. It is a serine protease that degrades elastin, a matrix component of tissues.

General Description

  • Elastase is a serine protease that can hydrolyze elastin, a specific protein of elastic fibers

Application

  • Elastase, porcine pancreas has been used in cell culture techniques, ELISA assays, and in experiments to measure the inhibitory activity of elastase
  • Since elastin is found in the fibers of connective tissues, elastase is commonly used to dissociate tissues that contain extensive intercellular fiber networks
  • It is typically used with other enzymes, including collagenase, trypsin, and chymotrypsin
  • Elastase enzyme is isolated from porcine pancreas and can be used alone or combined with other proteases for protein analysis by mass spectrometry and other applications, including membrane protein solubilization and protein sequence studies
  • This compound also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39445-21-1
Synonym E.C. 3.4.21.36; Pancreatopeptidase E

Specifications

For Use With (Application) A serine protease that acts on elastin
Content And Storage 2°C to 8°C
Form Lyophilized
Quantity 100 mg
Product Type Elastase

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.