Learn More
Elasticity of Gases Kit
Description
Includes
- Syringe cylinder
- Syringe cap
- Syringe piston
- Base
- Platen
- Lubricant
- Visual display of what happens when gases are subjected to increased pressure
- Plastic syringe with solid base and platen
- Add books, bricks, or other heavy objects to the platen to compress gas further
Specifications
Specifications
| For Use With (Application) | Ideal for gathering data in support of Boyle's law, students can plot pressure versus the reciprocal of the volume in search of a straight line |
| Includes | Syringe cylinder, syringe cap, syringe piston, base, platen, lubricant |
| Product Type | Elasticity of Gases Apparatus |