Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastin Antibody (ELN/1981), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275770AF405
Description
Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Elastin | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ELN | |
Recombinant full-length human Elastin protein (Uniprot: P15502) | |
0.1 mL | |
Extracellular Matrix | |
Elastin is a polymeric protein and is the main component of the extracellular matrix of arteries. It is synthesized and secreted as a soluble, single-chain protein (tropoelastin), which undergoes a number of post-translational modifications prior to its organization into an elastic fiber in the extracellular space. Elastin performs a regulatory function during arterial development by controlling proliferation of smooth muscle and stabilizing arterial structure. It imparts elasticity to the connective tissue. During aging, the elasticity of connective tissue is reduced because of the cross-linking of collagenous fibers with elastin. The abnormal accumulation of elastic tissue in blood vessels is found in atherosclerosis and hypertension. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ELN/1981 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction