Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Antibody (ELN/1981), DyLight 650, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP275770C

Catalog No. NB009061


Only null left
Add to Cart

Description

Description

Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Elastin
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
ELN
Recombinant full-length human Elastin protein (Uniprot: P15502)
0.1 mL
Extracellular Matrix
Elastin is a polymeric protein and is the main component of the extracellular matrix of arteries. It is synthesized and secreted as a soluble, single-chain protein (tropoelastin), which undergoes a number of post-translational modifications prior to its organization into an elastic fiber in the extracellular space. Elastin performs a regulatory function during arterial development by controlling proliferation of smooth muscle and stabilizing arterial structure. It imparts elasticity to the connective tissue. During aging, the elasticity of connective tissue is reduced because of the cross-linking of collagenous fibers with elastin. The abnormal accumulation of elastic tissue in blood vessels is found in atherosclerosis and hypertension.
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELN/1981
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
Mouse
Protein A or G purified
RUO
Primary
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.