Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Elastin Monoclonal Antibody (C3)

Catalog No. PIMA543843
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543843 20 μL
PIMA541583 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543843 Supplier Invitrogen™ Supplier No. MA543843
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Elastin Monoclonal Antibody for Western Blot

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elastin
Applications Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ELN
Gene Accession No. P15502
Gene Alias 5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS
Gene Symbols ELN
Host Species Mouse
Immunogen Recombinant protein Elastin. The antigen corresponds to amino acid range 392-645 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100628118, 2006
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.