Learn More
ELAVL1 Mouse anti-Human, Clone: OTI14F11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500872

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.Specifications
ELAVL1 | |
Monoclonal | |
Unconjugated | |
ELAVL1 | |
ELAV1, HUR, Hua, MelG | |
Mouse | |
Affinity Chromatography | |
RUO | |
1994 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI14F11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15717 | |
ELAVL1 | |
Full length human recombit protein of human ELAVL1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.