Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELAVL2/ELAVL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103196
Description
Antibody detects endogenous levels of total ELAV2/4.
Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.
Specifications
ELAVL2/ELAVL4 | |
Polyclonal | |
Unconjugated | |
ELAVL4 | |
Elav; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 2; ELAV like neuron-specific RNA binding protein 4; Elav2; ELAVL2; ELAVL3; Elavl4; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; ELAV-like protein 3; ELAV-like protein 4; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen D; HU-antigen B; Hu-antigen C; Hu-antigen D; HUB; HuC; HuD; mel-N1; nervous system-specific RNA-binding protein Hel-N1; Nervous system-specific RNA-binding protein Mel-N1; paraneoplastic encephalomyelitis antigen HuD; PLE21; PNEM; RGD1561943; r-HuD; RNA binding protein HuB; RNA-binding protein HUD3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15569, 15572, 1993, 1996, 286973, 432358 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O09032, P26378, Q12926, Q60899, Q61701, Q8CH84 | |
ELAVL2, ELAVL4 | |
A synthesized peptide derived from human ELAVL2(Accession Q12926), corresponding to amino acid residues E66-R116. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction