Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELAVL2 + ELAVL4 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981631
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1631 Supplier Bioss Supplier No. BS11743R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.

Specifications

Antigen ELAVL2 + ELAVL4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ELAVL2
Gene Accession No. O09032, P26378, Q12926, Q60899, Q61701, Q8CH84
Gene Alias Elav; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 2; ELAV like neuron-specific RNA binding protein 4; Elav2; ELAVL2; ELAVL3; ELAVL4; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; ELAV-like protein 3; ELAV-like protein 4; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen D; Hu-antigen B; Hu-antigen C; Hu-antigen D; HUB; HuC; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; Nervous system-specific RNA-binding protein Mel-N1; Paraneoplastic encephalomyelitis antigen HuD; PLE21; PNEM; RGD1561943; r-HuD; RNA binding protein HuB; RNA-binding protein HUD3
Gene Symbols ELAVL2, ELAVL4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ELAVL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15569, 15572, 1993, 1996, 286973, 432358
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.