Learn More
Invitrogen™ ELAVL2 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | ELAVL2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ELAVL2 |
| Gene Accession No. | Q12926, Q60899, Q8CH84 |
| Gene Alias | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV like neuron-specific RNA binding protein 2; Elav2; ELAVL 2; ELAVL 4; ELAVL2; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen B; Hu antigen D; hu-antigen B; HUB; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB; zgc:91918 |
| Gene Symbols | ELAVL2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.