Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELAVL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584578 Supplier Invitrogen™ Supplier No. PA584578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVRLCDVAS LLRSGSWAAE PWTGQVIAAM ETQLSNGPTC NNTANGPTTI NNNCSSPVDS GNTEDSKTNL Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

The protein encoded by this gene is a neural-specific RNA-binding protein that is known to bind to several 3' UTRs, including its own and also that of FOS and ID. The encoded protein may recognize a GAAA motif in the RNA. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAVL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELAVL2
Gene Accession No. Q12926
Gene Alias ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV like neuron-specific RNA binding protein 2; Elav2; ELAVL 2; ELAVL 4; ELAVL2; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen B; Hu antigen D; hu-antigen B; HUB; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB; zgc:91918
Gene Symbols ELAVL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELAVL2. Recombinant protein control fragment (Product #RP-103482).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.