Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELAVL3/ELAVL4 Mouse anti-Human, Mouse, Clone: 16A11 (Biotin), Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14090
Description
Mouse monoclonal antibody raised against human ELAVL3/ELAVL4.
- High specificity
- High reactivity
Specifications
ELAVL3/ELAVL4 | |
Monoclonal | |
Biotin | |
Immunofluorescence Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
Q14576;P26378 | |
ELAVL3 | |
Human ELAVL3/ELAVL4. | |
100 μg | |
Primary | |
Human, Mouse | |
Lyophilized |
Immunofluorescence, Western Blot | |
16A11 | |
Mouse monoclonal antibody raised against human ELAVL3/ELAVL4. | |
Lyophilized. | |
DKFZp547J036/HUC/HUCL/MGC20653/PLE21 | |
Mouse | |
Purified | |
RUO | |
1995/1996 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction